Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGCTCACTCTTCCCAGACCAC[C/G]CCCTCCTACAGGTGAGCACACCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609967 MIM: 604951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM129C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM129C - family with sequence similarity 129 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGLS - 6-phosphogluconolactonase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012088.2 | 1137 | Missense Mutation | CCC,GCC | P,A 163 | NP_036220.1 | |
XM_011527897.2 | 1137 | Missense Mutation | CCC,GCC | P,A 75 | XP_011526199.1 |