Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGCAGATGGACTGCTCCCCCAC[A/G]GCCAGCAGCAGTGCCAGTCCTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606700 | ||||||||||||||||||||
Literature Links: |
MIDN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIDN - midnolin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177401.4 | 1361 | Silent Mutation | ACA,ACG | T,T 239 | NP_796375.3 | |
XM_005259671.3 | 1361 | Silent Mutation | ACA,ACG | T,T 282 | XP_005259728.1 | |
XM_005259672.3 | 1361 | Silent Mutation | ACA,ACG | T,T 281 | XP_005259729.1 |