Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATGTTTGGTGAGATGTGAGTGC[C/T]GGTTAAACGCTCTGCCACAGTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF682 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF682 - zinc finger protein 682 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077349.1 | 1114 | Intron | NP_001070817.1 | |||
NM_033196.2 | 1114 | Intron | NP_149973.1 | |||
XM_017027455.1 | 1114 | Missense Mutation | CAG,CGG | Q,R 302 | XP_016882944.1 | |
XM_017027456.1 | 1114 | Intron | XP_016882945.1 |