Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTCCTGGGGGCACACCTCATT[A/G]GAGCCACTGAAGAGGGGCTCTCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604556 MIM: 134795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYRK1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYRK1B - dual specificity tyrosine phosphorylation regulated kinase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004714.2 | 1260 | Silent Mutation | TCC,TCT | S,S 316 | NP_004705.1 | |
NM_006483.2 | 1260 | Silent Mutation | TCC,TCT | S,S 316 | NP_006474.1 | |
NM_006484.2 | 1260 | Silent Mutation | TCC,TCT | S,S 316 | NP_006475.1 | |
XM_005259398.4 | 1260 | Silent Mutation | TCC,TCT | S,S 316 | XP_005259455.1 |
FBL - fibrillarin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6719 - microRNA 6719 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |