Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCGCTGAGAGCCCTGTTAATA[C/T]GTAAGACTGATATGGAAGGCAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLASRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLASRP - CLK4 associating serine/arginine rich protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278439.1 | 295 | Intron | NP_001265368.1 | |||
NM_007056.2 | 295 | Missense Mutation | ACG,ATG | T,M 66 | NP_008987.2 | |
XM_011526396.2 | 295 | Missense Mutation | ACG,ATG | T,M 66 | XP_011524698.1 | |
XM_011526397.2 | 295 | UTR 5 | XP_011524699.1 |