Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCTCCTCCCCAGCTCGCTGCTG[C/T]AAGGCCATGCCCTTCAGGAGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607335 MIM: 607824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEST2 - bestrophin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOOK2 - hook microtubule-tethering protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100176.1 | 2163 | Silent Mutation | TTA,TTG | L,L 672 | NP_001093646.1 | |
NM_013312.2 | 2163 | Silent Mutation | TTA,TTG | L,L 674 | NP_037444.2 | |
XM_005259873.2 | 2163 | Silent Mutation | TTA,TTG | L,L 673 | XP_005259930.1 | |
XM_005259874.2 | 2163 | Silent Mutation | TTA,TTG | L,L 671 | XP_005259931.1 | |
XM_005259876.4 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_005259933.1 | |
XM_005259877.4 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_005259934.1 | |
XM_011527943.2 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_011526245.1 | |
XM_011527944.2 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_011526246.1 | |
XM_011527945.2 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_011526247.1 | |
XM_011527946.2 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_011526248.1 | |
XM_011527947.2 | 2163 | Silent Mutation | TTA,TTG | L,L 539 | XP_011526249.1 | |
XM_017026720.1 | 2163 | Silent Mutation | TTA,TTG | L,L 601 | XP_016882209.1 | |
XM_017026721.1 | 2163 | Silent Mutation | TTA,TTG | L,L 599 | XP_016882210.1 |