Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGTGGGTTACAGGCCTCACAC[C/T]GCCCCGGGGATACTTCACTGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609694 MIM: 605110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMIP - GEM interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288998.1 | 2677 | Missense Mutation | AGT,GGT | S,G 847 | NP_001275927.1 | |
NM_001288999.1 | 2677 | Missense Mutation | AGT,GGT | S,G 850 | NP_001275928.1 | |
NM_016573.3 | 2677 | Missense Mutation | AGT,GGT | S,G 876 | NP_057657.2 | |
XM_005259927.2 | 2677 | Missense Mutation | AGT,GGT | S,G 873 | XP_005259984.1 | |
XM_011528048.2 | 2677 | Missense Mutation | AGT,GGT | S,G 755 | XP_011526350.1 | |
XM_017026861.1 | 2677 | UTR 3 | XP_016882350.1 | |||
XM_017026862.1 | 2677 | Intron | XP_016882351.1 |
LPAR2 - lysophosphatidic acid receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |