Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTACCCACTGGGCGCCATCGTGCG[C/T]TGTGACGCGGTGATGCCACCCGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614987 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS8L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPS8L1 - EPS8 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017729.3 | 422 | Intron | NP_060199.3 | |||
NM_133180.2 | 422 | Silent Mutation | CGC,CGT | R,R 106 | NP_573441.2 | |
XM_005259020.1 | 422 | Silent Mutation | CGC,CGT | R,R 106 | XP_005259077.1 | |
XM_011527050.1 | 422 | Intron | XP_011525352.1 | |||
XM_011527051.2 | 422 | Silent Mutation | CGC,CGT | R,R 106 | XP_011525353.1 | |
XM_011527052.2 | 422 | Silent Mutation | CGC,CGT | R,R 106 | XP_011525354.1 |