Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTGGCGGATGCCCATAGAGAAC[A/G]CTGGAGCCCTGGCTTTGTTCACGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODF3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ODF3L2 - outer dense fiber of sperm tails 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182577.2 | 576 | Missense Mutation | GCG,GTG | A,V 257 | NP_872383.1 | |
XM_005259545.4 | 576 | Missense Mutation | GCG,GTG | A,V 149 | XP_005259602.1 | |
XM_017026671.1 | 576 | Missense Mutation | GCG,GTG | A,V 149 | XP_016882160.1 |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |