Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTCTTCCTCACAGGCACGGCT[A/G]CCTGTCCTAATGGCAGCTTCCACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615956 MIM: 177060 | ||||||||||||||||||||
Literature Links: |
CCDC151 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC151 - coiled-coil domain containing 151 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKCSH - protein kinase C substrate 80K-H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001329.2 | 583 | Missense Mutation | ACC,GCC | T,A 69 | NP_001001329.1 | |
NM_001289102.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | NP_001276031.1 | |
NM_001289103.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | NP_001276032.1 | |
NM_001289104.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | NP_001276033.1 | |
NM_002743.3 | 583 | Missense Mutation | ACC,GCC | T,A 69 | NP_002734.2 | |
XM_011528130.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | XP_011526432.1 | |
XM_011528131.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | XP_011526433.1 | |
XM_011528132.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | XP_011526434.1 | |
XM_017026977.1 | 583 | Missense Mutation | ACC,GCC | T,A 69 | XP_016882466.1 |