Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTCCACCTACGACCGGGCCGG[C/T]GCTGTGCCCAGGACTCCGCTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605989 | ||||||||||||||||||||
Literature Links: |
PIAS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIAS4 - protein inhibitor of activated STAT 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015897.3 | 445 | Silent Mutation | GGC,GGT | G,G 94 | NP_056981.2 | |
XM_011528060.2 | 445 | Silent Mutation | GGC,GGT | G,G 113 | XP_011526362.1 | |
XM_017026868.1 | 445 | UTR 5 | XP_016882357.1 |