Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTTGGGCGACTTGAACTGCTC[C/G]TGGGAGCCTCTTGGGGACCTGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605350 MIM: 606855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL27RA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL27RA - interleukin 27 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004843.3 | 292 | Silent Mutation | TCC,TCG | S,S 53 | NP_004834.1 |
RLN3 - relaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311197.1 | 292 | Intron | NP_001298126.1 | |||
NM_080864.3 | 292 | Intron | NP_543140.1 |