Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCTTGGAGAAGCCGCCGCCAGG[A/C]GAGGGCGTCACGCTGTGGAAGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600637 | ||||||||||||||||||||
Literature Links: |
CCDC105 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC105 - coiled-coil domain containing 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173482.2 | 369 | Silent Mutation | GGA,GGC | G,G 90 | NP_775753.2 |
SLC1A6 - solute carrier family 1 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272087.1 | 369 | Intron | NP_001259016.1 | |||
NM_001272088.1 | 369 | Intron | NP_001259017.1 | |||
NM_005071.2 | 369 | Intron | NP_005062.1 | |||
XM_006722842.1 | 369 | Intron | XP_006722905.1 | |||
XM_006722844.1 | 369 | Intron | XP_006722907.1 | |||
XM_017027152.1 | 369 | Intron | XP_016882641.1 |