Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGTGACCCTGCTGTGTCAGTCA[C/T]GGAGCCCAATGGACACTTTTCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604821 | ||||||||||||||||||||
Literature Links: |
LILRB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LILRB4 - leukocyte immunoglobulin like receptor B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278426.3 | 678 | Missense Mutation | CGG,TGG | R,W 147 | NP_001265355.2 | |
NM_001278427.3 | 678 | Missense Mutation | CGG,TGG | R,W 147 | NP_001265356.2 | |
NM_001278428.3 | 678 | Missense Mutation | CGG,TGG | R,W 147 | NP_001265357.2 | |
NM_001278429.3 | 678 | Missense Mutation | CGG,TGG | R,W 176 | NP_001265358.2 | |
NM_001278430.3 | 678 | Intron | NP_001265359.2 | |||
XM_017026215.1 | 678 | Missense Mutation | CGG,TGG | R,W 188 | XP_016881704.1 | |
XM_017026216.1 | 678 | Missense Mutation | CGG,TGG | R,W 188 | XP_016881705.1 | |
XM_017026217.1 | 678 | Missense Mutation | CGG,TGG | R,W 188 | XP_016881706.1 |