Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAACAACAGGCTTTTAGTGTCGC[C/G]CCCCGCAGCCCCCGTCTACCCCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPE - lipase E, hormone sensitive type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005357.3 | 2363 | Missense Mutation | GCG,GGG | A,G 1074 | NP_005348.2 | |
XM_005258937.3 | 2363 | Missense Mutation | GCG,GGG | A,G 998 | XP_005258994.1 | |
XM_005258938.4 | 2363 | Missense Mutation | GCG,GGG | A,G 819 | XP_005258995.1 | |
XM_005258939.3 | 2363 | Missense Mutation | GCG,GGG | A,G 836 | XP_005258996.2 | |
XM_005258940.3 | 2363 | Missense Mutation | GCG,GGG | A,G 773 | XP_005258997.1 | |
XM_006723218.2 | 2363 | Missense Mutation | GCG,GGG | A,G 773 | XP_006723281.1 | |
XM_017026810.1 | 2363 | Missense Mutation | GCG,GGG | A,G 773 | XP_016882299.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101930071 - uncharacterized LOC101930071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |