Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCAGTAACACCGCCACGAAGA[A/C]GGGGACAAAGTGGGAGCTCCCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146631 MIM: 609950 | ||||||||||||||||||||
Literature Links: |
ICAM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ICAM3 - intercellular adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320605.1 | 2002 | Missense Mutation | GTC,TTC | V,F 393 | NP_001307534.1 | |
NM_001320606.1 | 2002 | Missense Mutation | GTC,TTC | V,F 412 | NP_001307535.1 | |
NM_001320608.1 | 2002 | Missense Mutation | GTC,TTC | V,F 228 | NP_001307537.1 | |
NM_002162.4 | 2002 | Missense Mutation | GTC,TTC | V,F 489 | NP_002153.2 |
RAVER1 - ribonucleoprotein, PTB binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133452.2 | 2002 | Intron | NP_597709.2 | |||
XM_011527671.1 | 2002 | Intron | XP_011525973.1 |