Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGTGCCATACAAACTGCCTGT[A/G]TCTTTGTCTGTTGGTTCCTGCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608717 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS13 - galectin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013268.2 | 76 | Silent Mutation | GTA,GTG | V,V 12 | NP_037400.1 | |
XM_011526874.2 | 76 | Intron | XP_011525176.1 | |||
XM_017026715.1 | 76 | Missense Mutation | ATC,GTC | I,V 48 | XP_016882204.1 |