Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACTGTCAGGTCCCACCTGTCCA[G/T]AGGTGGCCACCAGGACTTCTGGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615620 MIM: 601901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KPTN - kaptin, actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291296.1 | 928 | Missense Mutation | TAT,TCT | Y,S 275 | NP_001278225.1 | |
NM_007059.3 | 928 | Missense Mutation | TAT,TCT | Y,S 331 | NP_008990.2 | |
XM_011526399.2 | 928 | Missense Mutation | TAT,TCT | Y,S 393 | XP_011524701.1 | |
XM_017026226.1 | 928 | Missense Mutation | TAT,TCT | Y,S 415 | XP_016881715.1 | |
XM_017026227.1 | 928 | Missense Mutation | TAT,TCT | Y,S 415 | XP_016881716.1 | |
XM_017026228.1 | 928 | Missense Mutation | TAT,TCT | Y,S 415 | XP_016881717.1 | |
XM_017026229.1 | 928 | Intron | XP_016881718.1 |
NAPA-AS1 - NAPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC8A2 - solute carrier family 8 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |