Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCCCAGAGTGTCCACGTACC[A/G]CAGAAGTCGACATCATCGTACACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160994 MIM: 606466 | ||||||||||||||||||||
Literature Links: |
HNRNPM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPM - heterogeneous nuclear ribonucleoprotein M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAM1 - PML-RARA regulated adaptor molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032152.4 | 2043 | Silent Mutation | TGC,TGT | C,C 658 | NP_115528.4 | |
XM_005272502.2 | 2043 | Silent Mutation | TGC,TGT | C,C 658 | XP_005272559.1 | |
XM_011528352.2 | 2043 | Silent Mutation | TGC,TGT | C,C 660 | XP_011526654.1 | |
XM_011528353.2 | 2043 | Silent Mutation | TGC,TGT | C,C 660 | XP_011526655.1 |