Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGTCCCTAGGCTTGCCGCAGC[A/T]GATGGGTCTCATAGTCCTCTGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600310 MIM: 607536 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMP - cartilage oligomeric matrix protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000095.2 | 2294 | Missense Mutation | CAG,CTG | Q,L 753 | NP_000086.2 |
CRTC1 - CREB regulated transcription coactivator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098482.1 | 2294 | Intron | NP_001091952.1 | |||
NM_015321.2 | 2294 | Intron | NP_056136.2 | |||
XM_005259833.3 | 2294 | Intron | XP_005259890.1 | |||
XM_005259834.1 | 2294 | Intron | XP_005259891.1 | |||
XM_005259835.3 | 2294 | Intron | XP_005259892.1 | |||
XM_005259836.3 | 2294 | Intron | XP_005259893.1 | |||
XM_006722710.3 | 2294 | Intron | XP_006722773.1 | |||
XM_011527842.2 | 2294 | Intron | XP_011526144.1 | |||
XM_017026536.1 | 2294 | Intron | XP_016882025.1 | |||
XM_017026537.1 | 2294 | Intron | XP_016882026.1 | |||
XM_017026538.1 | 2294 | Intron | XP_016882027.1 |