Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACAGTGAGGATGCTGCAAAGC[C/T]GCACATAACTGTCTGACACCAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP9 - NLR family pyrin domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176820.3 | 694 | Silent Mutation | GCA,GCG | A,A 878 | NP_789790.2 | |
XM_011526894.2 | 694 | Silent Mutation | GCA,GCG | A,A 166 | XP_011525196.1 | |
XM_011526895.1 | 694 | Silent Mutation | GCA,GCG | A,A 166 | XP_011525197.1 |