Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCCTGCGCACAGCCTTTGAGT[C/T]GGGTGACCTCCCCCAGGCCGCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRGC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRGC - immunity related GTPase cinema | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019612.3 | 279 | Missense Mutation | TCG,TTG | S,L 33 | NP_062558.1 | |
XM_005259058.3 | 279 | Missense Mutation | TCG,TTG | S,L 70 | XP_005259115.1 | |
XM_006723283.3 | 279 | Missense Mutation | TCG,TTG | S,L 33 | XP_006723346.1 | |
XM_011527118.2 | 279 | Missense Mutation | TCG,TTG | S,L 33 | XP_011525420.1 |
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |