Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCGGACACTCAAGGCCTCCCTCC[C/T]TCGCAAGCGGGGCCGCCCCCCCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609133 | ||||||||||||||||||||
Literature Links: |
FIZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FIZ1 - FLT3 interacting zinc finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF524 - zinc finger protein 524 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153219.3 | 500 | Missense Mutation | CCT,CTT | P,L 51 | NP_694951.1 | |
XM_011526487.2 | 500 | Missense Mutation | CCT,CTT | P,L 157 | XP_011524789.1 |