Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCGGCCCTTCTGTTGGTACTTAC[C/T]TGAGATTTTCTGAGCCCATCCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC7A9 - solute carrier family 7 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126335.1 | 1516 | Missense Mutation | NP_001119807.1 | |||
NM_001243036.1 | 1516 | Missense Mutation | NP_001229965.1 | |||
NM_014270.4 | 1516 | Missense Mutation | NP_055085.1 | |||
XM_006722992.1 | 1516 | Missense Mutation | XP_006723055.1 | |||
XM_011526402.2 | 1516 | Missense Mutation | XP_011524704.1 | |||
XM_017026230.1 | 1516 | Missense Mutation | XP_016881719.1 |
TDRD12 - tudor domain containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |