Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCACGGCTGCTACGTTGGATTAT[C/T]GAAAGCAAAAGTTCCTAATCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSG11 - pregnancy specific beta-1-glycoprotein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113410.1 | 735 | Missense Mutation | AAT,GAT | N,D 210 | NP_001106881.1 | |
NM_002785.2 | 735 | Missense Mutation | AAT,GAT | N,D 332 | NP_002776.3 | |
NM_203287.1 | 735 | Missense Mutation | AAT,GAT | N,D 210 | NP_976032.2 |