Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCGCATCTTCTTCCTCTTTAA[C/G]GACATCCTGGTGTATGGCAGCATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615200 | ||||||||||||||||||||
Literature Links: |
PLEKHF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLEKHF1 - pleckstrin homology and FYVE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024310.4 | 279 | Missense Mutation | AAC,AAG | N,K 59 | NP_077286.3 | |
XM_005259256.3 | 279 | Missense Mutation | AAC,AAG | N,K 144 | XP_005259313.1 | |
XM_011527309.2 | 279 | Missense Mutation | AAC,AAG | N,K 59 | XP_011525611.1 |