Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCCTGGCGGCCCGTGGCCTCT[A/C]CTCTTTCCGCGGCCTCTCTTTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600957 MIM: 608743 MIM: 600796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMH - anti-Mullerian hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JSRP1 - junctional sarcoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144616.3 | 1279 | Missense Mutation | AAG,CAG | K,Q 265 | NP_653217.1 | |
XM_017026285.1 | 1279 | Nonsense Mutation | AAG,CAG | K,Q 318 | XP_016881774.1 | |
XM_017026286.1 | 1279 | Nonsense Mutation | AAG,CAG | K,Q 311 | XP_016881775.1 | |
XM_017026287.1 | 1279 | Nonsense Mutation | AAG,CAG | K,Q 272 | XP_016881776.1 |
MIR4321 - microRNA 4321 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SF3A2 - splicing factor 3a subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |