Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTCTGCTGCGGCTGCTGTCAT[C/T]GATCAAAGTGTGGGATGTGCTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606464 MIM: 159460 MIM: 600390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAMP - hepcidin antimicrobial peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021175.3 | 455 | Nonsense Mutation | CGA,TGA | R,* 75 | NP_066998.1 |
MAG - myelin associated glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF2 - upstream transcription factor 2, c-fos interacting | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |