Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCGCAGCAGCACCGGGATGAG[A/G]TCCTGTATGCGGCGGGCCGTTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615567 MIM: 604018 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ8B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ8B - coenzyme Q8B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142555.2 | 1706 | Silent Mutation | GAC,GAT | D,D 427 | NP_001136027.1 | |
NM_024876.3 | 1706 | Silent Mutation | GAC,GAT | D,D 468 | NP_079152.3 |
NUMBL - NUMB like, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |