Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGCCGTCTTCCTCCAGCGGCCC[A/G]TTCCGGGGATCCTCCTCGTCCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600040 MIM: 134790 MIM: 138570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAX - BCL2 associated X, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FTL - ferritin light chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GYS1 - glycogen synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161587.1 | 2236 | Silent Mutation | AAC,AAT | N,N 601 | NP_001155059.1 | |
NM_002103.4 | 2236 | Silent Mutation | AAC,AAT | N,N 665 | NP_002094.2 |