Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATTCCGGGCCACCAGGGCTGGC[G/T]GGCGCGAGGTGTAGTAGATTTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRL1 - adhesion G protein-coupled receptor L1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008701.2 | 4692 | Missense Mutation | CAG,CCG | Q,P 1428 | NP_001008701.1 | |
NM_014921.4 | 4692 | Missense Mutation | CAG,CCG | Q,P 1423 | NP_055736.2 | |
XM_005259818.3 | 4692 | Missense Mutation | CAG,CCG | Q,P 1447 | XP_005259875.1 | |
XM_011527796.2 | 4692 | Missense Mutation | CAG,CCG | Q,P 1459 | XP_011526098.1 | |
XM_011527798.2 | 4692 | Missense Mutation | CAG,CCG | Q,P 1454 | XP_011526100.1 | |
XM_011527801.2 | 4692 | Missense Mutation | CAG,CCG | Q,P 1379 | XP_011526103.1 | |
XM_017026475.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1460 | XP_016881964.1 | |
XM_017026476.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1460 | XP_016881965.1 | |
XM_017026477.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1453 | XP_016881966.1 | |
XM_017026478.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1436 | XP_016881967.1 | |
XM_017026479.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1430 | XP_016881968.1 | |
XM_017026480.1 | 4692 | Missense Mutation | CAG,CCG | Q,P 1374 | XP_016881969.1 | |
XM_017026481.1 | 4692 | Intron | XP_016881970.1 |
LOC100507373 - uncharacterized LOC100507373 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |