Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTACCCCAGCAAGTAGGTCAAA[C/T]TCAGGGTCAAAGAGCATGCAAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608792 MIM: 188070 | ||||||||||||||||||||
Literature Links: |
GIPC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIPC3 - GIPC PDZ domain containing family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXA2R - thromboxane A2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001060.5 | 1536 | UTR 3 | NP_001051.1 | |||
NM_201636.2 | 1536 | Silent Mutation | GAA,GAG | E,E 383 | NP_963998.2 | |
XM_011528214.2 | 1536 | Intron | XP_011526516.1 |