Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAACACACCCGAGGAGATCGCC[C/T]GCTTCCTGTACAAGGGCGAGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602488 MIM: 603953 | ||||||||||||||||||||
Literature Links: |
CYTH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYTH2 - cytohesin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004228.6 | 592 | Missense Mutation | CGC,TGC | R,C 98 | NP_004219.3 | |
NM_017457.5 | 592 | Missense Mutation | CGC,TGC | R,C 98 | NP_059431.1 | |
XM_006723472.2 | 592 | Missense Mutation | CGC,TGC | R,C 120 | XP_006723535.1 | |
XM_006723473.2 | 592 | Missense Mutation | CGC,TGC | R,C 120 | XP_006723536.1 |
KCNJ14 - potassium voltage-gated channel subfamily J member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372430 - uncharacterized LOC105372430 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |