Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCGCCAGTGAGATGTCCTGCG[A/G]CAGCTCGTCCTCGCTCTCCTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600006 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF15 - DDB1 and CUL4 associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PODNL1 - podocan like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX1 - regulatory factor X1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002918.4 | 3588 | Missense Mutation | CCG,TCG | P,S 938 | NP_002909.4 | |
XM_011528165.2 | 3588 | Missense Mutation | CCG,TCG | P,S 1048 | XP_011526467.1 | |
XM_011528166.2 | 3588 | Missense Mutation | CCG,TCG | P,S 1048 | XP_011526468.1 | |
XM_011528167.2 | 3588 | Missense Mutation | CCG,TCG | P,S 1048 | XP_011526469.1 | |
XM_011528168.2 | 3588 | Missense Mutation | CCG,TCG | P,S 950 | XP_011526470.1 | |
XM_011528169.2 | 3588 | Missense Mutation | CCG,TCG | P,S 939 | XP_011526471.1 | |
XM_011528170.2 | 3588 | Missense Mutation | CCG,TCG | P,S 939 | XP_011526472.1 |