Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCAGTGTGGACTCTCTGGTGT[A/G]TGTGAAAATGTGAGGCCTGACTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF235 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF235 - zinc finger protein 235 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004234.4 | 6258 | Missense Mutation | ACA,ATA | T,I 700 | NP_004225.3 | |
XM_005259412.4 | 6258 | Missense Mutation | ACA,ATA | T,I 696 | XP_005259469.1 |