Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGACCAGGCTGAGCGTGGTCTCT[C/T]GGTAGCGGGGGTGGCAGCTGACATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609694 MIM: 605110 MIM: 608127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMIP - GEM interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LPAR2 - lysophosphatidic acid receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004720.5 | 837 | Missense Mutation | CAA,CGA | Q,R 233 | NP_004711.2 | |
XM_011528421.2 | 837 | Missense Mutation | CAA,CGA | Q,R 233 | XP_011526723.1 | |
XM_017027468.1 | 837 | Missense Mutation | CAA,CGA | Q,R 233 | XP_016882957.1 | |
XM_017027469.1 | 837 | Missense Mutation | CAA,CGA | Q,R 233 | XP_016882958.1 |
PBX4 - PBX homeobox 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |