Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTTGGATGGGCAGGTGAGCACG[C/T]AGCCCAGGTGCTGGTTCCACATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123310 MIM: 606495 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKM - creatine kinase, M-type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001824.4 | 1011 | Missense Mutation | TAC,TGC | Y,C 279 | NP_001815.2 | |
XM_017026240.1 | 1011 | Missense Mutation | TAC,TGC | Y,C 279 | XP_016881729.1 |
MARK4 - microtubule affinity regulating kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |