Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCGAGGGAGTGTAGGCCAGGTG[A/G]GTGGGCGTCATGGGCAGAGCCGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611888 MIM: 606784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERF - ETS2 repressor factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301035.1 | 844 | Silent Mutation | ACC,ACT | T,T 198 | NP_001287964.1 | |
NM_001308402.1 | 844 | Silent Mutation | ACC,ACT | T,T 198 | NP_001295331.1 | |
NM_001312656.1 | 844 | Silent Mutation | ACC,ACT | T,T 198 | NP_001299585.1 | |
NM_006494.3 | 844 | Silent Mutation | ACC,ACT | T,T 273 | NP_006485.2 | |
XM_017026468.1 | 844 | Silent Mutation | ACC,ACT | T,T 198 | XP_016881957.1 | |
XM_017026469.1 | 844 | Silent Mutation | ACC,ACT | T,T 198 | XP_016881958.1 |
GSK3A - glycogen synthase kinase 3 alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |