Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCCCTTGCCCCCAGTACCTGC[A/G]AAGAACGTCATGAGCACTGCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154045 MIM: 606008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf84 - chromosome 19 open reading frame 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIM2 - lens intrinsic membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161748.1 | 626 | Silent Mutation | TTC,TTT | F,F 152 | NP_001155220.1 | |
NM_030657.3 | 626 | Silent Mutation | TTC,TTT | F,F 194 | NP_085915.2 |
NKG7 - natural killer cell granule protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |