Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGCACTCGGGGGAGCGCCCGTA[C/T]CAGTGCCCCATCTGCCGGCTGCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FIZ1 - FLT3 interacting zinc finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF524 - zinc finger protein 524 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153219.3 | 942 | Silent Mutation | TAC,TAT | Y,Y 198 | NP_694951.1 | |
XM_011526487.2 | 942 | Silent Mutation | TAC,TAT | Y,Y 304 | XP_011524789.1 |