Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTGAAACAGATGCTGCTGAACG[C/T]GGAGGTGAAGGCGGTGGGGCAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609828 MIM: 614715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSD1 - fibronectin type III and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024333.2 | 259 | Missense Mutation | GCG,GTG | A,V 36 | NP_077309.1 | |
XM_005259644.2 | 259 | Missense Mutation | GCG,GTG | A,V 36 | XP_005259701.1 |
TMIGD2 - transmembrane and immunoglobulin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |