Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCCCCTGCGGGCCCCCGATAGT[A/G]TTTGAGGAGGGGGTTTCCTTCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109190 | ||||||||||||||||||||
Literature Links: |
SLC1A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC1A5 - solute carrier family 1 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145144.1 | 1241 | Missense Mutation | CAC,TAC | H,Y 295 | NP_001138616.1 | |
NM_001145145.1 | 1241 | Missense Mutation | CAC,TAC | H,Y 321 | NP_001138617.1 | |
NM_005628.2 | 1241 | Missense Mutation | CAC,TAC | H,Y 523 | NP_005619.1 | |
XM_005259167.4 | 1241 | Intron | XP_005259224.1 |