Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTGCTGGGCTTCAGGCTCTGG[A/G]CCTCGGCTGTGTCATACTCCCGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606919 MIM: 602880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CERS1 - ceramide synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290265.1 | 1055 | Missense Mutation | GCC,GTC | A,V 230 | NP_001277194.1 | |
NM_021267.4 | 1055 | Missense Mutation | GCC,GTC | A,V 328 | NP_067090.1 | |
NM_198207.2 | 1055 | Missense Mutation | GCC,GTC | A,V 328 | NP_937850.1 |
GDF1 - growth differentiation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001492.5 | 1055 | UTR 5 | NP_001483.3 |