Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCCCCTGCAGTAGTGCTTCA[A/G]TGGGAATCAGATTGGACGGGGTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600172 MIM: 612276 | ||||||||||||||||||||
Literature Links: |
C1orf122 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf122 - chromosome 1 open reading frame 122 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTF1 - metal regulatory transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005955.2 | 2509 | Missense Mutation | ACT,ATT | T,I 735 | NP_005946.2 | |
XM_011541491.2 | 2509 | Missense Mutation | ACT,ATT | T,I 735 | XP_011539793.1 | |
XM_011541493.2 | 2509 | Intron | XP_011539795.1 | |||
XM_011541494.1 | 2509 | Missense Mutation | ACT,ATT | T,I 426 | XP_011539796.1 |
YRDC - yrdC N6-threonylcarbamoyltransferase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |