Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTACCGGAATGGGGACCCATT[C/T]TTCCCAGGCTCCCAGCTGGTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC28B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC28B - coiled-coil domain containing 28B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCDC2B - doublecortin domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099434.1 | 341 | Silent Mutation | TTC,TTT | F,F 20 | NP_001092904.1 | |
XM_017000413.1 | 341 | Silent Mutation | TTC,TTT | F,F 20 | XP_016855902.1 | |
XM_017000414.1 | 341 | Intron | XP_016855903.1 |
IQCC - IQ motif containing C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160042.1 | 341 | Intron | NP_001153514.1 | |||
NM_018134.2 | 341 | Intron | NP_060604.2 |