Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCAGGCCACAGCCCAGCAGCT[A/G]GTGCAGCTCCTCAGCAAGCAGAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611223 MIM: 613524 | ||||||||||||||||||||
Literature Links: |
AKT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKT3 - AKT serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206729.1 | 2039 | Intron | NP_001193658.1 | |||
NM_005465.4 | 2039 | Intron | NP_005456.1 | |||
NM_181690.2 | 2039 | Intron | NP_859029.1 | |||
XM_005272994.4 | 2039 | Intron | XP_005273051.1 | |||
XM_005272995.2 | 2039 | Intron | XP_005273052.1 | |||
XM_006711726.3 | 2039 | Intron | XP_006711789.1 | |||
XM_011544012.2 | 2039 | Intron | XP_011542314.1 | |||
XM_011544013.2 | 2039 | Intron | XP_011542315.1 | |||
XM_011544014.2 | 2039 | Intron | XP_011542316.1 | |||
XM_016999985.1 | 2039 | Intron | XP_016855474.1 |
SDCCAG8 - serologically defined colon cancer antigen 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006642.3 | 2039 | Silent Mutation | CTA,CTG | L,L 677 | NP_006633.1 | |
XM_005273013.4 | 2039 | Silent Mutation | CTA,CTG | L,L 634 | XP_005273070.1 | |
XM_005273018.1 | 2039 | Silent Mutation | CTA,CTG | L,L 536 | XP_005273075.1 | |
XM_005273021.4 | 2039 | Silent Mutation | CTA,CTG | L,L 376 | XP_005273078.1 | |
XM_005273022.3 | 2039 | Silent Mutation | CTA,CTG | L,L 370 | XP_005273079.1 | |
XM_005273023.4 | 2039 | Intron | XP_005273080.1 | |||
XM_006711727.3 | 2039 | Silent Mutation | CTA,CTG | L,L 687 | XP_006711790.1 | |
XM_006711728.3 | 2039 | Silent Mutation | CTA,CTG | L,L 644 | XP_006711791.1 | |
XM_011544021.2 | 2039 | Silent Mutation | CTA,CTG | L,L 719 | XP_011542323.1 | |
XM_011544022.2 | 2039 | Silent Mutation | CTA,CTG | L,L 709 | XP_011542324.1 | |
XM_011544023.2 | 2039 | Intron | XP_011542325.1 | |||
XM_011544024.2 | 2039 | Intron | XP_011542326.1 | |||
XM_011544026.2 | 2039 | Silent Mutation | CTA,CTG | L,L 640 | XP_011542328.1 | |
XM_011544028.2 | 2039 | Silent Mutation | CTA,CTG | L,L 565 | XP_011542330.1 | |
XM_011544029.2 | 2039 | Intron | XP_011542331.1 | |||
XM_011544030.2 | 2039 | Silent Mutation | CTA,CTG | L,L 362 | XP_011542332.1 | |
XM_017000103.1 | 2039 | Intron | XP_016855592.1 | |||
XM_017000104.1 | 2039 | Intron | XP_016855593.1 | |||
XM_017000105.1 | 2039 | Silent Mutation | CTA,CTG | L,L 598 | XP_016855594.1 | |
XM_017000106.1 | 2039 | Silent Mutation | CTA,CTG | L,L 579 | XP_016855595.1 | |
XM_017000107.1 | 2039 | Silent Mutation | CTA,CTG | L,L 376 | XP_016855596.1 | |
XM_017000108.1 | 2039 | Silent Mutation | CTA,CTG | L,L 376 | XP_016855597.1 | |
XM_017000109.1 | 2039 | Intron | XP_016855598.1 |