Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGCCAGGGTGTCCCCGGACCC[A/G]CCTTTCGGCCGGCAGATAGTCATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607868 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6742 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6742 - microRNA 6742 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM11 - tripartite motif containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145214.2 | 1632 | Silent Mutation | GGC,GGT | G,G 459 | NP_660215.1 | |
XM_011544285.2 | 1632 | Silent Mutation | GGC,GGT | G,G 382 | XP_011542587.1 | |
XM_017002412.1 | 1632 | Silent Mutation | GGC,GGT | G,G 458 | XP_016857901.1 |