Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGACAGTGAGTATGTGAATTT[C/T]GCCACCTGCTCCAGGTACAGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 191030 | ||||||||||||||||||||
Literature Links: |
C1orf189 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf189 - chromosome 1 open reading frame 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010979.2 | 138 | Missense Mutation | CAA,CGA | Q,R 27 | NP_001010979.1 |
C1orf43 - chromosome 1 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR190B - microRNA 190b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPM3 - tropomyosin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |