Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTCCAGCTCCACTGGCTCCATG[C/A]GGCTCAGTTTCTGCCTCAGCCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD34A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD34A - ankyrin repeat domain 34A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039888.3 | 2299 | Missense Mutation | NP_001034977.1 |
LIX1L - limb and CNS expressed 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3GL - RNA polymerase III subunit G like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005277431.4 | 2299 | Intron | XP_005277488.1 | |||
XM_011510059.2 | 2299 | Intron | XP_011508361.1 |