Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACTGGAAAATCTTTGTATAGG[G/T]GCTCCATTCATTTGACTCAAGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IVNS1ABP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IVNS1ABP - influenza virus NS1A binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006469.4 | 2391 | Missense Mutation | ACC,CCC | T,P 635 | NP_006460.2 | |
XM_005244843.3 | 2391 | Missense Mutation | ACC,CCC | T,P 417 | XP_005244900.1 | |
XM_011509079.2 | 2391 | Missense Mutation | ACC,CCC | T,P 642 | XP_011507381.1 | |
XM_011509080.2 | 2391 | Missense Mutation | ACC,CCC | T,P 553 | XP_011507382.1 | |
XM_017000084.1 | 2391 | Missense Mutation | ACC,CCC | T,P 424 | XP_016855573.1 |
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |